1-[2-(Aminomethyl)phenyl]-3-pyrrolidinol

Supplier: Matrix Scientific

052588-500MG 052588-2.5G
101900-238EA 364.62 USD
101900-238 101900-240
1-[2-(Aminomethyl)phenyl]-3-pyrrolidinol
1-[2-(Aminomethyl)phenyl]-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
MDL Number: MFCD13562318
CAS Number: 1220033-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR