[2-(4-Morpholinyl)phenyl]methanamine
Supplier: Matrix Scientific
Synonyms:
2-(Morpholin-4-yl)benzylamine
052587-500MG
052587-2.5G
101900-234EA
364.62
USD
101900-234
101900-236
[2-(4-Morpholinyl)phenyl]methanamine
[2-(4-Morpholinyl)phenyl]methanamine
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O MW: 192.26 g/mol |
MDL Number:
MFCD03086183 CAS Number: 204078-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...