2-[2-(Aminomethyl)(methyl)anilino]-1-ethanol

Supplier: Matrix Scientific

052585-500MG 052585-2.5G
101900-226EA 364.62 USD
101900-226 101900-228
2-[2-(Aminomethyl)(methyl)anilino]-1-ethanol
2-[2-(Aminomethyl)(methyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD12772571
CAS Number: 643087-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR