N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: Matrix Scientific

052583-500MG 052583-2.5G
101900-218EA 364.62 USD
101900-218 101900-220
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-Methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₃ClN₄O₂
MDL Number: MFCD13562317
CAS Number: 1220034-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR