2,3-Dihydro-1H-indol-1-yl-(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Supplier: Matrix Scientific

052580-500MG 052580-2.5G
101900-206EA 364.62 USD
101900-206 101900-208
2,3-Dihydro-1H-indol-1-yl-(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
2,3-Dihydro-1H-indol-1-yl-(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₇ClN₄O
MDL Number: MFCD13562314
CAS Number: 1220027-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR