3,4-Dihydro-2(1H)-isoquinolinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Supplier: Matrix Scientific

052579-500MG 052579-2.5G
101900-202EA 364.62 USD
101900-202 101900-204
3,4-Dihydro-2(1H)-isoquinolinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl
3,4-Dihydro-2(1H)-isoquinolinyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Hazard: Irritant

Formula: C₁₆H₁₉ClN₄O
MDL Number: MFCD13562313
CAS Number: 1219957-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR