[4-(2-Hydroxyethyl)-1-piperazinyl](4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Supplier: Matrix Scientific

052578-500MG 052578-2.5G
101900-198EA 364.62 USD
101900-198 101900-200
[4-(2-Hydroxyethyl)-1-piperazinyl](4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
[4-(2-Hydroxyethyl)-1-piperazinyl](4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₂ClN₅O₂
MDL Number: MFCD13562312
CAS Number: 1220027-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR