N-Cyclohexyl-N-ethyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl

Supplier: Matrix Scientific

052577-500MG 052577-2.5G
101900-194EA 364.62 USD
101900-194 101900-196
N-Cyclohexyl-N-ethyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl
N-Cyclohexyl-N-ethyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl

Hazard: Irritant

Formula: C₁₅H₂₅ClN₄O
MDL Number: MFCD13562311
CAS Number: 1219982-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR