(2-Ethyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Supplier: Matrix Scientific

052575-500MG 052575-2.5G
101900-186EA 364.62 USD
101900-186 101900-188
(2-Ethyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
(2-Ethyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₃ClN₄O
MW: 298.82 g/mol
MDL Number: MFCD13562309
CAS Number: 1220039-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR