N,N-Dipropyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: Matrix Scientific

052573-500MG 052573-2.5G
101900-178EA 364.62 USD
101900-178 101900-180
N,N-Dipropyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N,N-Dipropyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₃ClN₄O
MW: 286.81 g/mol
MDL Number: MFCD13562307
CAS Number: 1220034-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR