(4-Hydroxy-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Supplier: Matrix Scientific

052572-500MG 052572-2.5G
101900-174EA 364.62 USD
101900-174 101900-176
(4-Hydroxy-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
(4-Hydroxy-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₄O₂
MW: 286.76 g/mol
MDL Number: MFCD13562306
CAS Number: 1220033-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR