4-(4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-ylcarbonyl)-2-piperazinone hydrochloride

Supplier: Matrix Scientific

052571-500MG 052571-2.5G
101900-170EA 364.62 USD
101900-170 101900-172
4-(4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-ylcarbonyl)-2-piperazinone hydrochloride
4-(4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-ylcarbonyl)-2-piperazinone hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN₅O₂
MDL Number: MFCD13562305
CAS Number: 1220034-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR