1-Azepanyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
Supplier: Matrix Scientific
052570-500MG
052570-2.5G
101900-166EA
364.62
USD
101900-166
101900-168
1-Azepanyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
1-Azepanyl(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₁ClN₄O MW: 284.79 g/mol |
MDL Number:
MFCD13562304 CAS Number: 1219982-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...