(4-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Supplier: Matrix Scientific

052567-500MG 052567-2.5G
101900-154EA 364.62 USD
101900-154 101900-156
(4-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl
(4-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Hazard: Irritant

Formula: C₁₃H₂₁ClN₄O
MW: 284.79 g/mol
MDL Number: MFCD13562301
CAS Number: 1220029-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR