(3-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Supplier: Matrix Scientific

052566-500MG 052566-2.5G
101900-150EA 364.62 USD
101900-150 101900-152
(3-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride
(3-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₄O
MW: 284.79 g/mol
MDL Number: MFCD13562300
CAS Number: 1220035-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR