(2-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Supplier: Matrix Scientific

052565-500MG 052565-2.5G
101900-146EA 364.62 USD
101900-146 101900-148
(2-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl
(2-Methyl-1-piperidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Hazard: Irritant

Formula: C₁₃H₂₁ClN₄O
MW: 284.79 g/mol
MDL Number: MFCD13562299
CAS Number: 1220034-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR