N-Ethyl-N-(2-hydroxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
052563-500MG
052563-2.5G
101900-138EA
364.62
USD
101900-138
101900-140
N-Ethyl-N-(2-hydroxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-Ethyl-N-(2-hydroxyethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₉ClN₄O₂ |
MDL Number:
MFCD13562297 CAS Number: 1220034-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...