(3-Hydroxy-1-pyrrolidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Supplier: Matrix Scientific

052561-500MG 052561-2.5G
101900-130EA 364.62 USD
101900-130 101900-132
(3-Hydroxy-1-pyrrolidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl
(3-Hydroxy-1-pyrrolidinyl)(4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridin-3-yl)methanone HCl

Hazard: Irritant

Formula: C₁₁H₁₇ClN₄O₂
MDL Number: MFCD13562295
CAS Number: 1219976-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR