N-(2-Hydroxyethyl)-N-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Supplier: Matrix Scientific

052558-500MG 052558-2.5G
101900-118EA 364.62 USD
101900-118 101900-120
N-(2-Hydroxyethyl)-N-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(2-Hydroxyethyl)-N-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₇ClN₄O₂
MDL Number: MFCD13562292
CAS Number: 1219976-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR