4-(Ethylsulfonyl)-N1-methyl-N1-(tetrahydro-2H-pyran-4-ylmethyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052554-500MG 052554-2.5G
101900-102EA 364.62 USD
101900-102 101900-104
4-(Ethylsulfonyl)-N1-methyl-N1-(tetrahydro-2H-pyran-4-ylmethyl)-1,3-benzenediamine
4-(Ethylsulfonyl)-N1-methyl-N1-(tetrahydro-2H-pyran-4-ylmethyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₃S
MDL Number: MFCD13562288
CAS Number: 1220033-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR