N1-Benzyl-N1-ethyl-4-(ethylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052553-500MG 052553-2.5G
101900-098EA 364.62 USD
101900-098 101900-100
N1-Benzyl-N1-ethyl-4-(ethylsulfonyl)-1,3-benzenediamine
N1-Benzyl-N1-ethyl-4-(ethylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₂S
MDL Number: MFCD13562287
CAS Number: 1220039-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR