5-[3,4-Dihydro-1(2H)-quinolinyl]-2-(ethylsulfonyl)aniline

Supplier: Matrix Scientific

052552-500MG 052552-2.5G
101900-094EA 364.62 USD
101900-094 101900-096
5-[3,4-Dihydro-1(2H)-quinolinyl]-2-(ethylsulfonyl)aniline
5-[3,4-Dihydro-1(2H)-quinolinyl]-2-(ethylsulfonyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂S
MDL Number: MFCD13562286
CAS Number: 1220033-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR