N1-Cyclohexyl-N1-ethyl-4-(ethylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052548-500MG 052548-2.5G
101900-078EA 364.62 USD
101900-078 101900-080
N1-Cyclohexyl-N1-ethyl-4-(ethylsulfonyl)-1,3-benzenediamine
N1-Cyclohexyl-N1-ethyl-4-(ethylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₆H₂₆N₂O₂S
MDL Number: MFCD13562282
CAS Number: 1219972-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR