N1-Benzyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine

Supplier: Matrix Scientific

052547-500MG 052547-2.5G
101900-074EA 364.62 USD
101900-074 101900-076
N1-Benzyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine
N1-Benzyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₂S
MDL Number: MFCD13562281
CAS Number: 1220035-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR