N1-Cyclohexyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine

Supplier: Matrix Scientific

052545-500MG 052545-2.5G
101900-066EA 364.62 USD
101900-066 101900-068
N1-Cyclohexyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine
N1-Cyclohexyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₂S
MDL Number: MFCD13562279
CAS Number: 1220033-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR