1-[3-Amino-4-(ethylsulfonyl)phenyl]-4-piperidinol

Supplier: Matrix Scientific

052543-500MG 052543-2.5G
101900-058EA 364.62 USD
101900-058 101900-060
1-[3-Amino-4-(ethylsulfonyl)phenyl]-4-piperidinol
1-[3-Amino-4-(ethylsulfonyl)phenyl]-4-piperidinol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃S
MDL Number: MFCD13562277
CAS Number: 1220033-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR