4-[3-Amino-4-(ethylsulfonyl)phenyl]-2-piperazinone

Supplier: Matrix Scientific

052542-500MG 052542-2.5G
101900-054EA 364.62 USD
101900-054 101900-056
4-[3-Amino-4-(ethylsulfonyl)phenyl]-2-piperazinone
4-[3-Amino-4-(ethylsulfonyl)phenyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃S
MDL Number: MFCD13562276
CAS Number: 1220029-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR