5-(4-Benzyl-1-piperazinyl)-2-(ethylsulfonyl)phenylamine

Supplier: Matrix Scientific

052540-500MG 052540-2.5G
101900-046EA 364.62 USD
101900-046 101900-048
5-(4-Benzyl-1-piperazinyl)-2-(ethylsulfonyl)phenylamine
5-(4-Benzyl-1-piperazinyl)-2-(ethylsulfonyl)phenylamine

Hazard: Irritant

Formula: C₁₉H₂₅N₃O₂S
MW: 359.49 g/mol
MDL Number: MFCD13562274
CAS Number: 1219957-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR