2-(Ethylsulfonyl)-5-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

052536-500MG 052536-2.5G
101900-030EA 364.62 USD
101900-030 101900-032
2-(Ethylsulfonyl)-5-(2-methyl-1-piperidinyl)aniline
2-(Ethylsulfonyl)-5-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₂S
MW: 282.41 g/mol
MDL Number: MFCD13562270
CAS Number: 1220035-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR