N1-Butyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine

Supplier: Matrix Scientific

052533-500MG 052533-2.5G
101900-018EA 364.62 USD
101900-018 101900-020
N1-Butyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine
N1-Butyl-4-(ethylsulfonyl)-N1-methyl-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₂S
MDL Number: MFCD13562267
CAS Number: 1219957-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR