1-[3-Amino-4-(ethylsulfonyl)phenyl]-3-pyrrolidinol
Supplier: Matrix Scientific
052532-500MG
052532-2.5G
101900-014EA
364.62
USD
101900-014
101900-016
1-[3-Amino-4-(ethylsulfonyl)phenyl]-3-pyrrolidinol
1-[3-Amino-4-(ethylsulfonyl)phenyl]-3-pyrrolidinol
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₃S |
MDL Number:
MFCD13562266 CAS Number: 1220021-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...