2-(Ethylsulfonyl)-5-(4-morpholinyl)aniline

Supplier: Matrix Scientific

052531-500MG 052531-2.5G
101900-010EA 364.62 USD
101900-010 101900-012
2-(Ethylsulfonyl)-5-(4-morpholinyl)aniline
2-(Ethylsulfonyl)-5-(4-morpholinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃S
MDL Number: MFCD13562265
CAS Number: 1220034-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR