2-(Ethylsulfonyl)-5-(1-piperidinyl)aniline

Supplier: Matrix Scientific

052530-500MG 052530-2.5G
101900-006EA 364.62 USD
101900-006 101900-008
2-(Ethylsulfonyl)-5-(1-piperidinyl)aniline
2-(Ethylsulfonyl)-5-(1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD13562264
CAS Number: 1220021-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR