N1,N1-Diethyl-4-(ethylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052528-500MG 052528-2.5G
101899-998EA 364.62 USD
101899-998 101900-000
N1,N1-Diethyl-4-(ethylsulfonyl)-1,3-benzenediamine
N1,N1-Diethyl-4-(ethylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₂S
MDL Number: MFCD13562262
CAS Number: 1220028-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR