2-(Ethylsulfonyl)-5-(1-pyrrolidinyl)aniline

Supplier: Matrix Scientific

052527-500MG 052527-2.5G
101899-994EA 364.62 USD
101899-994 101899-996
2-(Ethylsulfonyl)-5-(1-pyrrolidinyl)aniline
2-(Ethylsulfonyl)-5-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂S
MDL Number: MFCD13562261
CAS Number: 1219972-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR