N1-Methyl-4-(methylsulfonyl)-N1-(tetrahydro-2H-pyran-4-ylmethyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052525-500MG 052525-2.5G
101899-986EA 364.62 USD
101899-986 101899-988
N1-Methyl-4-(methylsulfonyl)-N1-(tetrahydro-2H-pyran-4-ylmethyl)-1,3-benzenediamine
N1-Methyl-4-(methylsulfonyl)-N1-(tetrahydro-2H-pyran-4-ylmethyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₃S
MDL Number: MFCD13562259
CAS Number: 1219957-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR