N1-Benzyl-N1-ethyl-4-(methylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052524-500MG 052524-2.5G
101899-982EA 364.62 USD
101899-982 101899-984
N1-Benzyl-N1-ethyl-4-(methylsulfonyl)-1,3-benzenediamine
N1-Benzyl-N1-ethyl-4-(methylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₂S
MDL Number: MFCD13562258
CAS Number: 1220033-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR