5-[3,4-Dihydro-1(2H)-quinolinyl]-2-(methylsulfonyl)aniline

Supplier: Matrix Scientific

052523-500MG 052523-2.5G
101899-978EA 364.62 USD
101899-978 101899-980
5-[3,4-Dihydro-1(2H)-quinolinyl]-2-(methylsulfonyl)aniline
5-[3,4-Dihydro-1(2H)-quinolinyl]-2-(methylsulfonyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₂S
MDL Number: MFCD13562257
CAS Number: 1220039-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR