5-(2,3-Dihydro-1H-indol-1-yl)-2-(methylsulfonyl)phenylamine

Supplier: Matrix Scientific

052522-500MG 052522-2.5G
101899-974EA 364.62 USD
101899-974 101899-976
5-(2,3-Dihydro-1H-indol-1-yl)-2-(methylsulfonyl)phenylamine
5-(2,3-Dihydro-1H-indol-1-yl)-2-(methylsulfonyl)phenylamine

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂S
MDL Number: MFCD13562256
CAS Number: 1220033-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR