N1-Cyclohexyl-N1-ethyl-4-(methylsulfonyl)-1,3-benzenediamine
Supplier: Matrix Scientific
052519-500MG
052519-2.5G
101899-962EA
364.62
USD
101899-962
101899-964
N1-Cyclohexyl-N1-ethyl-4-(methylsulfonyl)-1,3-benzenediamine
N1-Cyclohexyl-N1-ethyl-4-(methylsulfonyl)-1,3-benzenediamine
Hazard: Irritant
Formula:
C₁₅H₂₄N₂O₂S |
MDL Number:
MFCD13562253 CAS Number: 1220017-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...