N1-Benzyl-N1-methyl-4-(methylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052518-500MG 052518-2.5G
101899-958EA 364.62 USD
101899-958 101899-960
N1-Benzyl-N1-methyl-4-(methylsulfonyl)-1,3-benzenediamine
N1-Benzyl-N1-methyl-4-(methylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂S
MDL Number: MFCD13562252
CAS Number: 1220028-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR