5-(2-Ethyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
Supplier: Matrix Scientific
052517-500MG
052517-2.5G
101899-954EA
364.62
USD
101899-954
101899-956
5-(2-Ethyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
5-(2-Ethyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
Hazard: Irritant
Formula:
C₁₄H₂₂N₂O₂S MW: 282.41 g/mol |
MDL Number:
MFCD13562251 CAS Number: 1220021-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...