4-[3-Amino-4-(methylsulfonyl)phenyl]-2-piperazinone

Supplier: Matrix Scientific

052513-500MG 052513-2.5G
101899-938EA 364.62 USD
101899-938 101899-940
4-[3-Amino-4-(methylsulfonyl)phenyl]-2-piperazinone
4-[3-Amino-4-(methylsulfonyl)phenyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₃S
MDL Number: MFCD13562247
CAS Number: 1220039-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR