5-(4-Benzyl-1-piperazinyl)-2-(methylsulfonyl)phenylamine

Supplier: Matrix Scientific

052512-500MG 052512-2.5G
101899-934EA 364.62 USD
101899-934 101899-936
5-(4-Benzyl-1-piperazinyl)-2-(methylsulfonyl)phenylamine
5-(4-Benzyl-1-piperazinyl)-2-(methylsulfonyl)phenylamine

Hazard: Irritant

Formula: C₁₈H₂₃N₃O₂S
MW: 345.47 g/mol
MDL Number: MFCD13562246
CAS Number: 1219972-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR