5-(4-Methyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine

Supplier: Matrix Scientific

052511-500MG 052511-2.5G
101899-930EA 364.62 USD
101899-930 101899-932
5-(4-Methyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine
5-(4-Methyl-1-piperidinyl)-2-(methylsulfonyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MW: 268.38 g/mol
MDL Number: MFCD13562245
CAS Number: 1220039-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR