N1,N1-Diallyl-4-(methylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052509-500MG 052509-2.5G
101899-922EA 364.62 USD
101899-922 101899-924
N1,N1-Diallyl-4-(methylsulfonyl)-1,3-benzenediamine
N1,N1-Diallyl-4-(methylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂S
MDL Number: MFCD13562243
CAS Number: 1220028-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR