2-[3-Amino(ethyl)-4-(methylsulfonyl)anilino]-1-ethanol

Supplier: Matrix Scientific

052508-500MG 052508-2.5G
101899-918EA 364.62 USD
101899-918 101899-920
2-[3-Amino(ethyl)-4-(methylsulfonyl)anilino]-1-ethanol
2-[3-Amino(ethyl)-4-(methylsulfonyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₈N₂O₃S
MDL Number: MFCD13562242
CAS Number: 1220017-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR