2-[3-Amino(methyl)-4-(methylsulfonyl)anilino]-1-ethanol

Supplier: Matrix Scientific

052505-500MG 052505-2.5G
101899-906EA 364.62 USD
101899-906 101899-908
2-[3-Amino(methyl)-4-(methylsulfonyl)anilino]-1-ethanol
2-[3-Amino(methyl)-4-(methylsulfonyl)anilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₃S
MDL Number: MFCD13562239
CAS Number: 1220028-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR