N1,N1-Dimethyl-4-(methylsulfonyl)-1,3-benzenediamine

Supplier: Matrix Scientific

052503-500MG 052503-2.5G
101899-898EA 364.62 USD
101899-898 101899-900
N1,N1-Dimethyl-4-(methylsulfonyl)-1,3-benzenediamine
N1,N1-Dimethyl-4-(methylsulfonyl)-1,3-benzenediamine

Hazard: Irritant

Formula: C₉H₁₄N₂O₂S
MDL Number: MFCD13562237
CAS Number: 1219976-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR