[4-(2-Hydroxyethyl)-1-piperazinyl](4-piperidinyl)methanone hydrochloride
Supplier: Matrix Scientific
052497-500MG
052497-2.5G
101899-874EA
364.62
USD
101899-874
101899-876
[4-(2-Hydroxyethyl)-1-piperazinyl](4-piperidinyl)methanone hydrochloride
[4-(2-Hydroxyethyl)-1-piperazinyl](4-piperidinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₄ClN₃O₂ MW: 277.8 g/mol |
MDL Number:
MFCD13562233 CAS Number: 1220028-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...